Methylenecyclohexane 98%

Danger

AC32551-0010 AC32551-0050
103532-452EA 229 USD
103532-452 200007-020
Methylenecyclohexane 98%
Methylenecyclohexane
Formula: C₇H₁₂
Boiling Pt: 102…103 °C (760 mmHg)
Melting Pt: –126 °C
Density: 0.8 g/cm³
Flash Pt: –006 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001661
CAS Number: 1192-37-6
EINECS: 203-624-3
UN: 1993
ADR: 3,II
Merck Index: 12,10412

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR