Acetonitrile-[D₃] 99.95 atom % D for NMR spectroscopy

Danger

Synonyms: Acetonitrile-d3Cyanomethane-D3Ethannitrile-D3Methyl-d3 cyanideAcetonitrile-[²H₃]Methyl cyanide-D3Trideuteroacetonitrile

AC32541-0100
200030-284EA 207.69 USD
200030-284
Acetonitrile-[D₃] 99.95 atom % D for NMR spectroscopy
Acetonitrile-[D3]

Packaged in 1.00 ml ampoules

Formula: D₃CCN
MW: 44.03 g/mol
Boiling Pt: 80.7 °C (1013 hPa)
Melting Pt: –46 °C
Density: 0.844 g/cm³ (20 °C)
Flash Pt: 2 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001881
CAS Number: 2206-26-0
EINECS: 218-616-5
UN: 1648
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR