Methanol-[D₄] 99.6+ atom % D for NMR spectroscopy
|
Danger
|
Synonyms:
Carbinol-D4
, Methyl alcohol-D4
, Methanol-[²H₄]
, Methyl-D3 alcohol-D
, Wood alcohol-D4
, Methanol-d4
, Hydroxy methane-D4
, Tetradeuteromethanol
AC32536-0100
AC32536-0500
200003-236EA
438
USD
200003-236
200003-234
Methanol-[D₄] 99.6+ atom % D for NMR spectroscopy
Methanol-[D4]
Formula:
D₃COD MW: 36.01 g/mol Boiling Pt: 65.3 °C (1013 hPa) Melting Pt: –99 °C Density: 0.888 g/cm³ (20 °C) Flash Pt: 12 °C Storage Temperature: Ambient |
MDL Number:
MFCD00044637 CAS Number: 811-98-3 EINECS: 212-378-6 UN: 1230 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...