Toluene-[D₈] 99.5+ atom % D for NMR spectroscopy

Danger

Synonyms: Toluene-[²H₈]Phenylmethane-D8Methylbenzene-D8

AC32132-0010 AC32132-0100
200007-906EA 30.58 USD
200007-906 200020-968
Toluene-[D₈] 99.5+ atom % D for NMR spectroscopy
Toluene-[D8]
Formula: C₆D₅CD₃
MW: 100.08 g/mol
Boiling Pt: 110 °C (1013 hPa)
Melting Pt: –84 °C
Density: 0.94 g/cm³ (20 °C)
Flash Pt: 4 °C (closed cup)
Storage Temperature: Refrigerator
MDL Number: MFCD00044638
CAS Number: 2037-26-5
EINECS: 218-009-5
UN: 1294
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR