1,1,2,2-Tetrachloroethane-[D₂] (99.5% D) for NMR spectroscopy
|
Danger
|
Synonyms:
1,1,2,2-Tetrachloroethane-[²H₂]
AC32131-0010
AC32131-0250
103527-612EA
184
USD
103527-612
200008-358
1,1,2,2-Tetrachloroethane-[D₂] (99.5% D) for NMR spectroscopy
1,1,2,2-Tetrachloroethane-[D2]
Formula:
Cl₂CDCDCl₂ MW: 169.83 g/mol Boiling Pt: 146 °C (760 mmHg) Density: 1.61 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00037672 CAS Number: 33685-54-0 EINECS: 251-634-1 UN: 1702 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...