Deuterium oxide 99.9 atom % D for NMR spectroscopy

Synonyms: Heavy waterWater-D2

AC32071-0075
200055-412EA 42.69 USD
200055-412
Deuterium oxide 99.9 atom % D for NMR spectroscopy
Deuterium oxide

Packaged in 0.75 ml ampoules

Formula: D₂O
MW: 20 g/mol
Boiling Pt: 101.3 °C (1013 hPa)
Melting Pt: 3.8 °C
Density: 1.105 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00044636
CAS Number: 7789-20-0
EINECS: 232-148-9
Merck Index: 12,02984

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR