Bromocyclobutane 97%, pure

Danger

Synonyms: Cyclobutyl bromide

AC32050-0250 AC32050-0010 AC32050-0050
103532-444EA 200 USD
103532-444 103532-446 200011-812
Bromocyclobutane 97%, pure
Bromocyclobutane
Formula: C₄H₇Br
MW: 135 g/mol
Boiling Pt: 108 °C
Density: 1.43 g/cm³
Flash Pt: 22 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001317
CAS Number: 4399-47-7
EINECS: 224-530-9
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR