L(-)-Glutathione (oxidised form) ≥98%

Supplier: ACROS ORGANICS MS

Synonyms: L(-)-Glutathione, oxidizedGSSGBi(glutathion-S-yl)Glutathione oxidizedGlutathiolOxidized Glutathione (GSSG)

AC32022-0050 AC32022-5000
103512-348EA 191.68 USD
103512-348 200003-194
L(-)-Glutathione (oxidised form) ≥98%
L(-)-Glutathione (oxidized form)
Formula: C₂₀H₃₂N₆O₁₂S₂
MW: 612.64 g/mol
Melting Pt: 185 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063106
CAS Number: 27025-41-8
EINECS: 248-170-7
UN: 0000

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤8 % (105°C, 3 h)
HPLC ≥97.5 %
Residue after ignition ≤0.5 %
Specific optical rotation -92° to -106° (20°C, 589 nm) (c=4, water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR