2-Amino-4-methyl-3-nitropyridine 98%

Warning

Synonyms: 4-Methyl-3-nitro-2-pyridylamine4-Methyl-3-nitropyridin-2-amine2-Amino-3-nitro-4-picoline

AC31955-0050 AC31955-0010 AC31955-0250
103532-440EA 213 USD
103532-440 200046-660 200016-298
2-Amino-4-methyl-3-nitropyridine 98%
2-Amino-4-methyl-3-nitropyridine
Formula: C₆H₇N₃O₂
MW: 153.14 g/mol
Melting Pt: 136…141 °C
Storage Temperature: Ambient
MDL Number: MFCD00006315
CAS Number: 6635-86-5
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR