4,4-Dimethyl-2-cyclohexen-1-one 97%

Warning

Synonyms: 4,4-Dimethylcyclohex-2-enone

AC31954-0250 AC31954-0050
103512-328EA 239 USD
103512-328 200003-192
4,4-Dimethyl-2-cyclohexen-1-one 97%
4,4-Dimethyl-2-cyclohexen-1-one
Formula: C₈H₁₂O
MW: 124.18 g/mol
Boiling Pt: 72…73.5 °C (20 mmHg)
Density: 0.94 g/cm³
Flash Pt: 64 °C
Storage Temperature: Ambient
MDL Number: MFCD00009695
CAS Number: 1073-13-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR