Samarium trinitrate hexahydrate 99.9% (trace metals basis)

Danger

Synonyms: Samarium (III) nitrate hexahydrateSamarium(III) nitrate hydrate

AC31615-1000
103532-404EA 198 USD
103532-404
Samarium trinitrate hexahydrate 99.9% (trace metals basis)
Samarium trinitrate hexahydrate
Formula: N₃O₉Sm·6H₂O
MW: 444.47 g/mol
Melting Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00149857
CAS Number: 13759-83-6
EINECS: 233-798-6
UN: 1477
ADR: 5.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR