N-Methylethylamine 97%

Danger

Synonyms: N-Ethylmethylamine

AC31468-0050 AC31468-0250
200013-632EA 70.2 USD
200013-632 103527-528
N-Methylethylamine 97%
N-Methylethylamine
Formula: C₃H₉N
MW: 59.11 g/mol
Boiling Pt: 33…34 °C (760 mmHg)
Density: 0.68 g/cm³
Flash Pt: <–34 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009030
CAS Number: 624-78-2
EINECS: 210-862-1
UN: 2733
ADR: 3,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR