Benzoyl isothiocyanate 98%

Danger

AC31463-0050 AC31463-0500
103532-384EA 226 USD
103532-384 200011-348
Benzoyl isothiocyanate 98%
Benzoyl isothiocyanate
Formula: C₈H₅NOS
MW: 163.2 g/mol
Boiling Pt: 128…131 °C (15 mmHg)
Melting Pt: –1 °C
Density: 1.21 g/cm³
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004815
CAS Number: 532-55-8
EINECS: 208-540-0
UN: 2810
ADR: 6.1,III
Merck Index: 13,01115

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR