3-Chloropropyl isocyanate 96%

Danger

AC31459-0010 AC31459-0050
200029-256EA 113.5 USD
200029-256 103532-382
3-Chloropropyl isocyanate 96%
3-Chloropropyl isocyanate
Formula: C₄H₆ClNO
MW: 119.55 g/mol
Boiling Pt: 152…154 °C
Density: 1.165 g/cm³
Flash Pt: 62 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010384
CAS Number: 13010-19-0
UN: 2206
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR