Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Fluoro-6-iodobenzoic acid ≥98%

Supplier: ACROS ORGANICS MS

Warning

AC31194-0010
200022-658EA 68.76 USD InStock
200022-658
2-Fluoro-6-iodobenzoic acid ≥98%
2-Fluoro-6-iodobenzoic acid


Formula: C₇H₄FIO₂
MW: 266.01 g/mol
Boiling Pt: 312 °C (1013 hPa)
Melting Pt: 123…126 °C
MDL Number: MFCD00042289
CAS Number: 111771-08-5
EINECS: 000-000-0
Order Now


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty