2-Fluoro-4-iodoaniline 98%

Warning

AC31193-0050 AC31193-0250
200054-146EA 92.1 USD
200054-146 200054-148
2-Fluoro-4-iodoaniline 98%
2-Fluoro-4-iodoaniline
Formula: C₆H₅FIN
MW: 237.02 g/mol
Boiling Pt: 259 °C (1013 hPa)
Melting Pt: 51…57 °C
Flash Pt: 113 °C
MDL Number: MFCD00011738
CAS Number: 29632-74-4
UN: 2811

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR