2,4-Bis(trifluoromethyl)benzoic acid 98%

Warning

AC31173-0010
200016-074EA 30.9 USD
200016-074
2,4-Bis(trifluoromethyl)benzoic acid 98%
2,4-Bis(trifluoromethyl)benzoic acid
Formula: C₉H₄F₆O₂
MW: 258.12 g/mol
Boiling Pt: 260 °C (1013 hPa)
Melting Pt: 109…113 °C
MDL Number: MFCD00042496
CAS Number: 32890-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR