2,6-Difluorophenol 98%

Danger

AC31163-0250 AC31163-0050
103532-322EA 246 USD
103532-322 200028-386
2,6-Difluorophenol 98%
2,6-Difluorophenol
Formula: C₆H₄F₂O
MW: 130.09 g/mol
Boiling Pt: 59…61 °C (17 mmHg)
Melting Pt: 38…42 °C
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00002158
CAS Number: 28177-48-2
EINECS: 248-884-9
UN: 2925
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR