3,5-Difluorocinnamic acid 98%

Warning

AC31159-0010
200047-324EA 38.1 USD
200047-324
3,5-Difluorocinnamic acid 98%
3,5-Difluorocinnamic acid
Formula: C₉H₆F₂O₂
MW: 184.14 g/mol
Melting Pt: 203…206 °C
MDL Number: MFCD00010321
CAS Number: 84315-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR