Allylpentafluorobenzene 97%

Warning

Synonyms: 1-Allyl-2,3,4,5,6-pentafluorobenzene3-(Pentafluorophenyl)-1-propene

AC31085-0250 AC31085-0050
103532-308EA 148.3 USD
103532-308 200024-328
Allylpentafluorobenzene 97%
Allylpentafluorobenzene
Formula: (C₆F₅)CH₂CHCH₂
MW: 208.13 g/mol
Boiling Pt: 148…149 °C
Melting Pt: –64 °C
Density: 1.358 g/cm³
Flash Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD00000301
CAS Number: 1736-60-3
EINECS: 217-083-6
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR