3-Methylbiphenyl 95%

Synonyms: 3-Phenyltoluene

AC30984-0010
200005-304EA 65.5 USD
200005-304
3-Methylbiphenyl 95%
3-Methylbiphenyl
Formula: C₁₃H₁₂
MW: 168.24 g/mol
Boiling Pt: 272 °C
Melting Pt: 4…5 °C
Density: 1.01 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008533
CAS Number: 643-93-6
EINECS: 211-404-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR