4-Methoxyphenylboronic acid ≥97%

Supplier: ACROS ORGANICS MS

Warning

Synonyms: (4-Methoxyphenyl)metaboric acidDihydroxy(4-methoxyphenyl)borane4-Methoxybenzeneboronic acid

AC30948-0050 AC30948-0010 AC30948-0250
103511-484EA 80.27 USD InStock
103511-484 200003-156 103511-486
4-Methoxyphenylboronic acid ≥97%
4-Methoxyphenylboronic acid
Formula: C₇H₉BO₃
MW: 151.96 g/mol
Melting Pt: 204 - 207 °C
Storage Temperature: Ambient
MDL Number: MFCD00039139
CAS Number: 5720-07-0
EINECS: 000-000-0

Order Now

Specification Test Results

Appearance White to light brown Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR