5-Iodo-m-xylene 99%

Warning

Synonyms: 1-Iodo-3,5-dimethylbenzene3,5-Dimethyliodobenzene1,3-Dimethyl-5-iodobenzene3,5-Dimethyl-1-iodobenzene

AC30925-0250 AC30925-0050
200042-312EA 35.8 USD
200042-312 200010-740
5-Iodo-m-xylene 99%
5-Iodo-m-xylene
Formula: C₈H₉I
MW: 232.06 g/mol
Boiling Pt: 92…94 °C (3 mmHg)
Density: 1.6 g/cm³
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00060659
CAS Number: 22445-41-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR