Ethyl-3-bromobenzoate 98%

Warning

Synonyms: 3-Bromobenzoic acid ethyl esterEthyl 3-bromobenzoate

AC30854-1000 AC30854-0050 AC30854-0250
200049-344EA 94.6 USD
200049-344 103512-272 200049-346
Ethyl-3-bromobenzoate 98%
Ethyl 3-bromobenzoate
Formula: C₉H₉BrO₂
MW: 229.07 g/mol
Boiling Pt: 130…131 °C (12 mmHg)
Density: 1.43 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00013529
CAS Number: 24398-88-7
EINECS: 246-223-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR