3-Hexyne 99%

Danger

AC30812-0250 AC30812-0050
200009-948EA 100 USD
200009-948 103532-258
3-Hexyne 99%
3-Hexyne
Formula: C₆H₁₀
MW: 82.15 g/mol
Boiling Pt: 81…82 °C
Melting Pt: –61.5 °C
Density: 0.72 g/cm³
Flash Pt: –14 °C
Storage Temperature: Ambient
MDL Number: MFCD00009381
CAS Number: 928-49-4
EINECS: 213-173-4
UN: 3295
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR