2,6-Difluoropyridine 99%

Warning

AC30666-0050 AC30666-0250
200022-326EA 38.6 USD
200022-326 103532-246
2,6-Difluoropyridine 99%
2,6-Difluoropyridine
Formula: C₅H₃F₂N
MW: 115.08 g/mol
Boiling Pt: 124.5 °C (743 mmHg)
Density: 1.26 g/cm³
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00006227
CAS Number: 1513-65-1
EINECS: 216-152-8
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR