1-Bromo-3,3-dimethylbutanone 93%
|
Warning
|
AC30641-0050
AC30641-0250
200008-618EA
41.9
USD
200008-618
103512-246
1-Bromo-3,3-dimethylbutanone 93%
1-Bromo-3,3-dimethylbutanone
Formula:
C₆H₁₁BrO MW: 179.06 g/mol Boiling Pt: 189…192 °C (760 mmHg) Melting Pt: –10 °C Density: 1.33 g/cm³ Flash Pt: 75 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000206 CAS Number: 5469-26-1 EINECS: 226-794-0 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...