α,α,α-Trifluoro-4-iodotoluene 97%
|
Warning
|
Synonyms:
1-Iodo-4-(trifluoromethyl)benzene
, 4-Iodobenzotrifluoride
, α,α,α-Trifluoro-p-iodotoluene
AC30618-0050
AC30618-0010
200005-468EA
81.3
USD
200005-468
200054-692
α,α,α-Trifluoro-4-iodotoluene 97%
α,α,α-Trifluoro-4-iodotoluene
Formula:
C₇H₄F₃I MW: 272.01 g/mol Boiling Pt: 76…79 °C (15 mmHg) Melting Pt: 17 °C Density: 1.851 g/cm³ Flash Pt: 68 °C |
MDL Number:
MFCD00039398 CAS Number: 455-13-0 EINECS: 207-234-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...