1,1-Bis(methylthio)-2-nitroethylene 99%

Warning

AC30569-0050 AC30569-0250
200033-628EA 44 USD
200033-628 103512-242
1,1-Bis(methylthio)-2-nitroethylene 99%
1,1-Bis(methylthio)-2-nitroethylene
Formula: C₄H₇NO₂S₂
MW: 165.24 g/mol
Melting Pt: 124…126 °C
Storage Temperature: Ambient
MDL Number: MFCD00010443
CAS Number: 13623-94-4
EINECS: 237-108-4
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR