4,6-Dihydroxy-5-nitropyrimidine ≥95%

Warning

Synonyms: 5-Nitropyrimidine-4,6-diol6-Hydroxy-5-nitro-1H-pyrimidin-4-one4,6-Dihydroxy-5-nitropyrimdine

AC30541-0050 AC30541-0250
200063-618EA 23 USD
200063-618 103512-236
4,6-Dihydroxy-5-nitropyrimidine ≥95%
4,6-Dihydroxy-5-nitropyrimidine
Formula: C₄H₃N₃O₄
MW: 157.09 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00006110
CAS Number: 2164-83-2
EINECS: 218-504-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR