1-Bromo-2-ethylbutane 97%

Warning

Synonyms: 3-(Bromomethyl)pentane

AC30518-0050
200033-876EA 71.8 USD
200033-876
1-Bromo-2-ethylbutane 97%
1-Bromo-2-ethylbutane
Formula: C₆H₁₃Br
MW: 165.07 g/mol
Boiling Pt: 143…144 °C (760 mmHg)
Density: 1.17 g/cm³
Flash Pt: 35 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000219
CAS Number: 3814-34-4
EINECS: 242-659-9
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR