Cycloheptene 95%

Danger

AC30426-0250 AC30426-0050
200042-996EA 305 USD
200042-996 200018-606
Cycloheptene 95%
Cycloheptene
Formula: C₇H₁₂
Boiling Pt: 114.6 °C (1013 hPa)
Melting Pt: –56 °C
Density: 0.82 g/cm³
Flash Pt: –006 °C
MDL Number: MFCD00004156
CAS Number: 628-92-2
EINECS: 211-060-4
UN: 2242
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR