1-Pentyne 99%

Danger

AC30382-0050 AC30382-1000 AC30382-0250
103532-202EA 28.8 USD
103532-202 200008-766 103532-204
1-Pentyne 99%
1-Pentyne
Formula: C₅H₈
MW: 68.12 g/mol
Boiling Pt: 40 °C
Melting Pt: 106…105 °C
Density: 0.69 g/cm³
Flash Pt: <–34 °C
Storage Temperature: Ambient
MDL Number: MFCD00009469
CAS Number: 627-19-0
EINECS: 210-987-1
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR