α-Bromo-2,6-difluorotoluene 97%

Danger

Synonyms: 2-(Bromomethyl)-1,3-difluorobenzene2,6-Difluorobenzyl bromide

AC30357-0250 AC30357-0050
200024-362EA 56 USD
200024-362 103532-196
α-Bromo-2,6-difluorotoluene 97%
α-Bromo-2,6-difluorotoluene
Formula: C₇H₅BrF₂
MW: 207.02 g/mol
Melting Pt: 51…55 °C
Flash Pt: >109 °C
Storage Temperature: Ambient
MDL Number: MFCD00000329
CAS Number: 85118-00-9
EINECS: 285-652-6
UN: 1759
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR