2,5-Difluorobenzaldehyde 98%

Warning

AC30352-0010 AC30352-0050 AC30352-0250
103532-194EA 189 USD
103532-194 103532-192 200009-024
2,5-Difluorobenzaldehyde 98%
2,5-Difluorobenzaldehyde
Formula: C₇H₄F₂O
MW: 142.1 g/mol
Boiling Pt: 67…69 °C (17 mmHg)
Density: 1.3 g/cm³
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00010327
CAS Number: 2646-90-4
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR