2,3-Difluorobenzaldehyde 98%

Warning

AC30351-0010 AC30351-0050
200028-140EA 32.7 USD
200028-140 103532-190
2,3-Difluorobenzaldehyde 98%
2,3-Difluorobenzaldehyde
Formula: C₇H₄F₂O
MW: 142.1 g/mol
Boiling Pt: 64…65 °C (17 mmHg)
Density: 1.3 g/cm³
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00010292
CAS Number: 2646-91-5
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR