1-Amino-2,2,2-trifluoroethane 99.5%

Danger

AC30350-0500 AC30350-0010 AC30350-0100
103527-454EA 1337 USD
103527-454 200003-132 200003-130
1-Amino-2,2,2-trifluoroethane 99.5%
1-Amino-2,2,2-trifluoroethane
Formula: C₂H₄F₃N
MW: 99.06 g/mol
Boiling Pt: 37…38 °C (760 mmHg)
Density: 1.24 g/cm³
Flash Pt: –16 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008132
CAS Number: 753-90-2
EINECS: 212-041-3
UN: 3286
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR