1-Octene 99+%

Danger

AC30125-0010 AC30125-1000 AC30125-5000 AC30125-0100 AC30125-0025
102777-344EA 294 USD
102777-344 200008-756 103527-432 103527-434 200003-112
1-Octene 99+%
1-Octene
Formula: C₈H₁₆
MW: 112.22 g/mol
Boiling Pt: 121…123 °C (760 mmHg)
Melting Pt: –102 °C
Density: 0.715 g/cm³
Flash Pt: 13 °C
Storage Temperature: Ambient
MDL Number: MFCD00009548
CAS Number: 111-66-0
EINECS: 203-893-7
UN: 3295
ADR: 3,II
Merck Index: 13,01770

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR