cis-1,2-Cyclohexanediol 99%
Synonyms:
cis-Cyclohexane-1,2-diol
AC30097-0050
AC30097-0010
103511-466EA
110
USD
103511-466
200011-684
cis-1,2-Cyclohexanediol 99%
cis-1,2-Cyclohexanediol
Formula:
C₆H₁₂O₂ Melting Pt: 98…101 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064944 CAS Number: 1792-81-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...