Ethanol-[D₆], anhydrous (99% D) for NMR spectroscopy

Supplier: ACROS ORGANICS MS

Danger

Synonyms: Hydroxy ethane-D6Ethanol-[²H₆]Ethyl alcohol-D6Methyl carbinol-D6

AC30068-0010 AC30068-0050
200025-392EA 56.63 USD InStock
200025-392 103527-426
Ethanol-[D₆], anhydrous (99% D) for NMR spectroscopy
Ethanol-[D6]
Formula: D₃CCD₂OD
MW: 52.02 g/mol
Boiling Pt: 78 °C (1013 hPa)
Melting Pt: -114,5 °C
Density: 0.801 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00051020
CAS Number: 1516-08-1
EINECS: 216-162-2
UN: 1170
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
D-Enrichment ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR