Vanadium(IV) chloride 99+%

Danger

Synonyms: Vanadium tetrachloride

AC29969-1000
103527-414EA 172 USD
103527-414
Vanadium(IV) chloride 99+%
Vanadium (IV) chloride
Formula: Cl₄V
MW: 192.75 g/mol
Boiling Pt: 153 °C (760 mmHg)
Melting Pt: –20.5 °C
Density: 1.82 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00061343
CAS Number: 7632-51-1
EINECS: 231-561-1
UN: 2444
ADR: 8,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR