1,2-Dibromo-1-phenylethane 97%

Danger

Synonyms: (1,2-Dibromoethyl)benzene

AC29930-5000
200033-230EA 160 USD
200033-230
1,2-Dibromo-1-phenylethane 97%
1,2-Dibromo-1-phenylethane
Formula: C₆H₅CHBrCH₂Br
MW: 263.96 g/mol
Boiling Pt: 139…141 °C (15 mmHg)
Melting Pt: 68…73 °C
Storage Temperature: Ambient
MDL Number: MFCD00000138
CAS Number: 93-52-7
EINECS: 202-253-4
UN: 3261
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/07/83/760783.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR