2-Phenylquinoline 99+%

Warning

AC29799-0010 AC29799-0050
200022-908EA 77.4 USD
200022-908 103464-894
2-Phenylquinoline 99+%
2-Phenylquinoline
Formula: C₁₅H₁₁N
MW: 205.26 g/mol
Boiling Pt: 363 °C (760 mmHg)
Melting Pt: 82…84 °C
Storage Temperature: Ambient
MDL Number: MFCD00011568
CAS Number: 612-96-4
EINECS: 210-326-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR