Cyclopentene 95+%

Danger

AC29590-0025 AC29590-5000 AC29590-1000
103527-390EA 672 USD
103527-390 200018-628 103527-392
Cyclopentene 95+%
Cyclopentene
Formula: C₅H₈
Boiling Pt: 44 °C (760 mmHg)
Melting Pt: –135 °C
Density: 0.774 g/cm³
Flash Pt: –30 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001394
CAS Number: 142-29-0
EINECS: 205-532-9
UN: 2246
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR