4-Vinylbenzoic acid 96%

Warning

AC29491-2500 AC29491-0010 AC29491-0050
103512-142EA 23.8 USD
103512-142 200017-726 103512-140
4-Vinylbenzoic acid 96%
4-Vinylbenzoic acid
Formula: C₉H₈O₂
MW: 148.16 g/mol
Melting Pt: 142…144 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002569
CAS Number: 1075-49-6
EINECS: 214-053-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR