4-Fluoroanthranilic acid 96%

Warning

Synonyms: 2-Amino-4-fluorobenzoic acid

AC29302-0025 AC29302-0100
103512-102EA 284 USD
103512-102 200003-038
4-Fluoroanthranilic acid 96%
4-Fluoroanthranilic acid
Formula: C₇H₆FNO₂
MW: 155.13 g/mol
Melting Pt: 188…196 °C
Storage Temperature: Ambient
MDL Number: MFCD00075553
CAS Number: 446-32-2
EINECS: 207-163-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR