2-Adamantanone 98%

Synonyms: Adamantan-2-one

AC29250-1000 AC29250-0050 AC29250-0250
103512-100EA 370 USD
103512-100 200046-620 200009-112
2-Adamantanone 98%
2-Adamantanone
Formula: C₁₀H₁₄O
MW: 150.22 g/mol
Melting Pt: 282…286 °C
Storage Temperature: Ambient
MDL Number: MFCD00074737
CAS Number: 700-58-3
EINECS: 211-847-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR