Indane 95%

Warning

Synonyms: 2,3-Dihydro-1H-indeneIndan

AC29248-5000 AC29248-0050 AC29248-1000
200025-562EA 63.5 USD
200025-562 200031-568 200049-668
Indane 95%
Indane
Formula: C₉H₁₀
MW: 118.18 g/mol
Boiling Pt: 176.5 °C (1013 hPa)
Melting Pt: –51 °C
Density: 0.96 g/cm³
Flash Pt: 50 °C
Storage Temperature: Ambient
MDL Number: MFCD00003795
CAS Number: 496-11-7
EINECS: 207-814-7
UN: 1993
ADR: 3,III
Merck Index: 13,04956

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR