4,4-Dimethoxybutanone 92%

Warning

Synonyms: Acetylacetaldehyde dimethyl acetalAcetoacetaldehyde-1,1-Dimethyl Acetal4,4-Dimethoxybutan-2-one3-Oxobutyraldehyde dimethyl acetal4,4-Dimethoxy-2-butanone

AC29233-0010 AC29233-2500
200063-572EA 281 USD
200063-572 200017-988
4,4-Dimethoxybutanone 92%
4,4-Dimethoxybutanone
Formula: C₆H₁₂O₃
MW: 132.16 g/mol
Boiling Pt: 178 °C (1013 hPa)
Melting Pt: –82 °C
Density: 0.99 g/cm³
Flash Pt: 49 °C
MDL Number: MFCD00008789
CAS Number: 5436-21-5
EINECS: 226-605-1
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR