4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane (Kryptofix® 222) 98%

Warning

Synonyms: Fludeoxyglucose Related Compound A4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo(8.8.8)hexacosane4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosaneCryptand 222(2.2.2)CryptandKryptofix 222

AC29195-0010
200003-030EA 170 USD
200003-030
4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane (Kryptofix® 222) 98%
4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane (Kryptofix® 222)
Formula: C₁₈H₃₆N₂O₆
MW: 376.49 g/mol
Melting Pt: 68…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00005111
CAS Number: 23978-09-8
EINECS: 245-962-4
UN: 2811
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/14/35/20961435.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR